Learn More
Cayman Chemical ML-324, Purity: ≥95%, CAS: 1222800-79-4, 1 mg
SDP
Catalog No. 502966107
Encompass_Preferred
A cell-permeable inhibitor of JMJD2E (KDM4DL; IC50 = 920 nM); reduces the expression of immediate early genes of HSV (IC50 = ~10 µM) and hCMV, blocks viral infection, and suppresses the level of viral reactivation in a ganglia explant model of latently infected mice
Catalog No. 50-296-6107 Supplier Cayman Chemical Supplier No. 174721 MG
A cell-permeable inhibitor of JMJD2E (KDM4DL; IC50 = 920 nM); reduces the expression of immediate early genes of HSV (IC50 = ~10 µM) and hCMV, blocks viral infection, and suppresses the level of viral reactivation in a ganglia explant model of latently infected mice