Learn More
Cayman Chemical ML-335, Purity: ≥98%, CAS: 825658-06-8, 5 mg
SDP
Catalog No. 502939943
Encompass_Preferred
An activator of K2P2.1/TREK1 and K2P10.1/TREK2 (EC50s = 14.3 and 5.2 µM, respectively, in Xenopus oocytes)
Catalog No. 50-293-9943 Supplier Cayman Chemical Supplier No. 276355 MG
An activator of K2P2.1/TREK1 and K2P10.1/TREK2 (EC50s = 14.3 and 5.2 µM, respectively, in Xenopus oocytes)