Learn More
Cayman Chemical ML-792, Purity: ≥98%, CAS: 1644342-14-2, 5 mg
SDP
Catalog No. 502909731
Encompass_Preferred
An inhibitor of SUMO-activating enzyme (IC50s = 0.003 and 0.011 µM with SUMO1 or SUMO2, respectively, as the ubiquitin-like proteins); selective for SAE over NAE and UAE (IC50s = 32 and >100 µM, respectively); decreases global SUMOylation in HCT116 cells (EC50 = 0.019 µM), as well as decreases the viability of MDA-MB-468 and A375 cells (EC50s = 0.06 and 0.45 µM, respectively); reduces tumor growth in an HCT116 mouse xenograft model at 150 and 200 mg/kg twice per day
Catalog No. 50-290-9731 Supplier Cayman Chemical Supplier No. 368205 MG
An inhibitor of SUMO-activating enzyme (IC50s = 0.003 and 0.011 µM with SUMO1 or SUMO2, respectively, as the ubiquitin-like proteins); selective for SAE over NAE and UAE (IC50s = 32 and >100 µM, respectively); decreases global SUMOylation in HCT116 cells (EC50 = 0.019 µM), as well as decreases the viability of MDA-MB-468 and A375 cells (EC50s = 0.06 and 0.45 µM, respectively); reduces tumor growth in an HCT116 mouse xenograft model at 150 and 200 mg/kg twice per day