Learn More
Cayman Chemical ANTIBODY ML-SA5 10mg
SDP
Catalog No. 502916770
Encompass_Preferred
A TRPML1 agonist; activates whole-endolysosome IML1 in vacuolin-1-enlarged isolated vacuoles (EC50 = 333 nM); is cytotoxic to MeWo and M12 melanoma cells but not normal melanocytes at 3 µM; induces gastric acid secretion in isolated stomach from wild-type but not TRPML1-/- mice; decreases the necrotic area and percentage of centrally nucleated fibers in the dystrophin-deficient mdx mouse model of Duchenne muscular dystrophy at 2 and 5 mg/kg
Catalog No. 50-291-6770 Supplier Cayman Chemical Supplier No. 3832610 MG
A TRPML1 agonist; activates whole-endolysosome IML1 in vacuolin-1-enlarged isolated vacuoles (EC50 = 333 nM); is cytotoxic to MeWo and M12 melanoma cells but not normal melanocytes at 3 µM; induces gastric acid secretion in isolated stomach from wild-type but not TRPML1-/- mice; decreases the necrotic area and percentage of centrally nucleated fibers in the dystrophin-deficient mdx mouse model of Duchenne muscular dystrophy at 2 and 5 mg/kg