Learn More
Cayman Chemical MLN0905, Purity: ≥98%, CAS: 1228960-69-7, 5 mg
SDP
Catalog No. 502990335
Encompass_Preferred
A Plk1 inhibitor (IC50 = 2 nM); inhibits 95% of Plk1 activity but also inhibits the activity of 10 additional kinases by greater than or equal to 90% in a panel of 359 kinases at 1 µM; reduces H3S10Ph in HT-29 colorectal cancer cells (EC30 = 9 nM); inhibits proliferation in a panel of six human lymphoma cell lines (IC50 = 3.2-24 nM)
Catalog No. 50-299-0335 Supplier Cayman Chemical Supplier No. 311155 MG
A Plk1 inhibitor (IC50 = 2 nM); inhibits 95% of Plk1 activity but also inhibits the activity of 10 additional kinases by greater than or equal to 90% in a panel of 359 kinases at 1 µM; reduces H3S10Ph in HT-29 colorectal cancer cells (EC30 = 9 nM); inhibits proliferation in a panel of six human lymphoma cell lines (IC50 = 3.2-24 nM)