Learn More
Cayman Chemical MLN4760, Purity: ≥98%, CAS: 305335-31-3, 25 mg
SDP
Catalog No. 502990721
Encompass_Preferred
An ACE2 inhibitor (IC50 = 0.44 nM); selective for ACE2 over ACE and CPDA (IC50s = >100 and 27 µM, respectively); inhibits ACE2 hydrolysis of angiotensin I in vitro at 1 µM; inhibits the protein-protein interaction between ACE2 and the SARS-CoV-2 spike glycoprotein; inhibits exogenous recombinant ACE2-induced prevention of angiotensin II-induced hypertension in mice
Catalog No. 50-299-0721 Supplier Cayman Chemical Supplier No. 3132725 MG
An ACE2 inhibitor (IC50 = 0.44 nM); selective for ACE2 over ACE and CPDA (IC50s = >100 and 27 µM, respectively); inhibits ACE2 hydrolysis of angiotensin I in vitro at 1 µM; inhibits the protein-protein interaction between ACE2 and the SARS-CoV-2 spike glycoprotein; inhibits exogenous recombinant ACE2-induced prevention of angiotensin II-induced hypertension in mice