Learn More
Cayman Chemical MLN8054, Purity: ≥98%, CAS: 869363-13-3, 1 mg
SDP
Catalog No. 502937331
Encompass_Preferred
An inhibitor of Aurora A kinase (IC50 = 4 nM); selective for Aurora A over Aurora B, PKC, CaMKII, Cdk2E, CK2, LCK, PKA, Chk1, Chk2, Cdk1, and Plk1 (IC50s = 0.172-100 µM); induces cell cycle arrest at the G2/M phase and the formation of abnormal mitotic spindles in HCT116 colorectal and PC3 prostate cancer cells; inhibits the growth of various cancer cells (IC50s = 0.11-1.43 µM); reduces tumor volume in HCT116 and PC3 mouse xenograft models at 30 mg/kg
Catalog No. 50-293-7331 Supplier Cayman Chemical Supplier No. 273401 MG
An inhibitor of Aurora A kinase (IC50 = 4 nM); selective for Aurora A over Aurora B, PKC, CaMKII, Cdk2E, CK2, LCK, PKA, Chk1, Chk2, Cdk1, and Plk1 (IC50s = 0.172-100 µM); induces cell cycle arrest at the G2/M phase and the formation of abnormal mitotic spindles in HCT116 colorectal and PC3 prostate cancer cells; inhibits the growth of various cancer cells (IC50s = 0.11-1.43 µM); reduces tumor volume in HCT116 and PC3 mouse xenograft models at 30 mg/kg