Learn More
Cayman Chemical MLS-573151, Purity: ≥98%, CAS: 10179-57-4, 1 mg
SDP
Catalog No. 502969467
Encompass_Preferred
A specific, cell permeable inhibitor of Cdc42 (IC50 = 2 μM); has no effect on other GTPases, including the related Rho family members RhoA, Rac1, and Rac2
Catalog No. 50-296-9467 Supplier Cayman Chemical Supplier No. 152691 MG
A specific, cell permeable inhibitor of Cdc42 (IC50 = 2 μM); has no effect on other GTPases, including the related Rho family members RhoA, Rac1, and Rac2