Learn More
Cayman Chemical MNS, Purity: ≥95%, CAS: 1485-00-3, 100 mg
SDP
Catalog No. 502971798
Encompass_Preferred
Blocks and reverses platelet aggregation induced by such triggers as collagen, U-46619, and ADP (IC50 = 1.1, 2.1, and 4.1 µM, respectively); reduces tyrosine phosphorylation in stimulated platelets and inhibits recombinant human Syk and Src (IC50 = 2.8 and 27.3 µM, respectively) in vitro
Catalog No. 50-297-1798 Supplier Cayman Chemical Supplier No. 15206100 MG
Blocks and reverses platelet aggregation induced by such triggers as collagen, U-46619, and ADP (IC50 = 1.1, 2.1, and 4.1 µM, respectively); reduces tyrosine phosphorylation in stimulated platelets and inhibits recombinant human Syk and Src (IC50 = 2.8 and 27.3 µM, respectively) in vitro