Learn More
Cayman Chemical ANTIBODY Mometason 500ug
SDP
Catalog No. 502992244
Encompass_Preferred
A synthetic glucocorticoid; reduces pulmonary edema and neutrophil infiltration in a mouse model of chlorine-induced chemical lung injury at 3 mg/kg; intranasal administration reduces nose-rubbing behavior, sneezing, and nasal mucosal levels of IL-4 in a mouse model of allergic rhinitis at 0.2 UG/animal; reduces nasal mucosal epithelial destruction, fibrosis, and cilia loss in a rat model of oxymetazoline-induced rhinitis medicamentosa
Catalog No. 50-299-2244 Supplier Cayman Chemical Supplier No. 31702500 UG
A synthetic glucocorticoid; reduces pulmonary edema and neutrophil infiltration in a mouse model of chlorine-induced chemical lung injury at 3 mg/kg; intranasal administration reduces nose-rubbing behavior, sneezing, and nasal mucosal levels of IL-4 in a mouse model of allergic rhinitis at 0.2 UG/animal; reduces nasal mucosal epithelial destruction, fibrosis, and cilia loss in a rat model of oxymetazoline-induced rhinitis medicamentosa