Learn More
Cayman Chemical Motolimod, Purity: ≥98%, CAS: 926927-61-9, 5 mg
SDP
Catalog No. 502964481
Encompass_Preferred
A TLR8 agonist; increases the production of TNF-α and IL-12 in human PBMCs (EC50s = 140 and 120 nM, respectively), monocytes, and dendritic cells; increases cytolysis in K562 NK cell-sensitive leukemia cells at 167 and 500 nM for 48 hours; increases plasma levels of a group of human cytokines , including IL-6, IL-12p70, TNF-α, MCP-1, and MIP-1β, in NSG-HIS mice at 1.5 and 15 mg/m2; reduces tumor growth and increases tumor infiltration of monocytes and T cells in an ovarian cancer NSG-HIS mouse model
Catalog No. 50-296-4481 Supplier Cayman Chemical Supplier No. 229525 MG
A TLR8 agonist; increases the production of TNF-α and IL-12 in human PBMCs (EC50s = 140 and 120 nM, respectively), monocytes, and dendritic cells; increases cytolysis in K562 NK cell-sensitive leukemia cells at 167 and 500 nM for 48 hours; increases plasma levels of a group of human cytokines , including IL-6, IL-12p70, TNF-α, MCP-1, and MIP-1β, in NSG-HIS mice at 1.5 and 15 mg/m2; reduces tumor growth and increases tumor infiltration of monocytes and T cells in an ovarian cancer NSG-HIS mouse model