Learn More
Cayman Chemical Moxonidine, Purity: ≥95%, CAS: 75438-57-2, 25 mg
SDP
Catalog No. 502936690
Encompass_Preferred
An imidazoline I1 and α2A-AR agonist (Kis = 56 and 150 nM, respectively, in radioligand binding assays); selective for α2A-AR and I1 receptors over α1-, α2B, and α2C-ARs (Kis = >30,000, 1,000, and 2,000 nM, respectively); decreases mean arterial pressure in spontaneously hypertensive rats at 0.2 nmol injected into the RVLM; reduces blood pressure in cynomolgus monkeys at 167 µg/kg
Catalog No. 50-293-6690 Supplier Cayman Chemical Supplier No. 2563925 MG
An imidazoline I1 and α2A-AR agonist (Kis = 56 and 150 nM, respectively, in radioligand binding assays); selective for α2A-AR and I1 receptors over α1-, α2B, and α2C-ARs (Kis = >30,000, 1,000, and 2,000 nM, respectively); decreases mean arterial pressure in spontaneously hypertensive rats at 0.2 nmol injected into the RVLM; reduces blood pressure in cynomolgus monkeys at 167 µg/kg