Learn More
Cayman Chemical ANTIBODY MPLA 100ug
SDP
Catalog No. 502936276
Encompass_Preferred
A synthetic derivative of lipid A; a TLR4 agonist; increases injection site transcription of MyD88- and TRIF-dependent genes as well as genes encoding cytokines and cytokine receptors, chemokines, and molecules involved in the complement pathway and antigen presentation at 5 μg/100 μl, i.m.; increases the number of CD3+ T cells and CD11c+ GR1+ dendritic cells recruited to draining lymph nodes; increases serum levels of CCL2/MCP-1, CXCL1, CSF3, IL-6, and IL-10 by more than 10-fold compared to control animals; emulsions containing MPLA have demonstrated efficacy as vaccine adjuvants in several mouse models
Catalog No. 50-293-6276 Supplier Cayman Chemical Supplier No. 25998100 UG
A synthetic derivative of lipid A; a TLR4 agonist; increases injection site transcription of MyD88- and TRIF-dependent genes as well as genes encoding cytokines and cytokine receptors, chemokines, and molecules involved in the complement pathway and antigen presentation at 5 μg/100 μl, i.m.; increases the number of CD3+ T cells and CD11c+ GR1+ dendritic cells recruited to draining lymph nodes; increases serum levels of CCL2/MCP-1, CXCL1, CSF3, IL-6, and IL-10 by more than 10-fold compared to control animals; emulsions containing MPLA have demonstrated efficacy as vaccine adjuvants in several mouse models