Learn More
Cayman Chemical MPP-PICA, Purity: ≥98%, 1 mg
SDP
Catalog No. 502990055
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-299-0055 Supplier Cayman Chemical Supplier No. 312681 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications