Learn More
Cell Biolabs Inc Mouse Anti-Methylglyoxal Monoclonal Antibody, clone 3D11, 100 µg
Supplier:  Cell Biolabs Inc STA011
Encompass_Preferred
Methylglyoxal (MG) is formed through non-oxidative mechanisms from triose phosphates during anaerobic glycolysis and it can modify amino acids, nucleic acids, and proteins. MG reacts with arginine, lysine and cysteine residues of proteins to form AGEs. MG is involved in various pathological processes. For example, MG derivatives are found elevated in diabetes. Cell Biolabs' Mouse Anti-Methylglyoxal Monoclonal Antibody is Affinity purified and suitable for Western blot or Immunohistochemistry. The antibody will work with protein samples from any species. 100 ug at a concentration of 0.6 mg/mL.
Catalog No. NC0281426