Learn More
Cayman Chemical ANTIBODY MSD-M1PAM 500ug
SDP
Supplier:  Cayman Chemical 41358500 UG
Encompass_Preferred
A positive allosteric modulator of M1 mAChRs; enhances ACh-induced calcium production in a calcium mobilization assay from 0.1-10 µM; reverses scopolamine-induced deficits in freezing behavior in mice from 1-30 mg/kg; attenuates deficits in self-ordered spatial search in cynomolgus macaques and in the object retrieval detour task in rhesus macaques
Catalog No. 50-294-5762