Learn More
Cayman Chemical MSX-122, Purity: ≥98%, CAS: 897657-95-3, 5 mg
SDP
Catalog No. 502986563
Encompass_Preferred
A partial antagonist of CXCR4; reduces inflammation in a mouse model of experimental colitis; prevents the production of TNF-α, IL-1β, and IL-6; reduces TNF-α secretion by macrophages infected with patient-derived invasive E. coli; decreases metastases in three different mouse models
Catalog No. 50-298-6563 Supplier Cayman Chemical Supplier No. 234265 MG
A partial antagonist of CXCR4; reduces inflammation in a mouse model of experimental colitis; prevents the production of TNF-α, IL-1β, and IL-6; reduces TNF-α secretion by macrophages infected with patient-derived invasive E. coli; decreases metastases in three different mouse models