Learn More
Cayman Chemical MY-5445, Purity: ≥98%, CAS: 78351-75-4, 10 mg
SDP
Catalog No. 502983478
Encompass_Preferred
A selective inhibitor of PDE5 (Ki = 1.3 µM); relaxes intact strips of rat aorta (EC50 = 4 µM)
Catalog No. 50-298-3478 Supplier Cayman Chemical Supplier No. 2087210 MG
A selective inhibitor of PDE5 (Ki = 1.3 µM); relaxes intact strips of rat aorta (EC50 = 4 µM)