Learn More
Cayman Chemical ANTIBODY MZ1 500ug
SDP
Supplier:  Cayman Chemical 21622500 UG
Encompass_Preferred
A PROTAC that drives the selective proteasomal degradation of BRD4; also causes the removal of BRD2 and BRD3 when used at higher concentrations
Catalog No. 50-298-7579