Learn More
Cayman Chemical N6022, Purity: ≥98%, CAS: 1208315-24-5, 25 mg
SDP
Catalog No. 502981453
Encompass_Preferred
A tight-binding, specific, and fully reversible inhibitor of GSNOR (IC50 = 8 nM; Ki = 2.5 nM)
Catalog No. 50-298-1453 Supplier Cayman Chemical Supplier No. 2126925 MG
A tight-binding, specific, and fully reversible inhibitor of GSNOR (IC50 = 8 nM; Ki = 2.5 nM)