Learn More
Cayman Chemical NAP-1, Purity: ≥98%, CAS: 131721-28-3, 5 mg
SDP
Catalog No. 502937134
Encompass_Preferred
A compound with anesthetic activity; increases paired-pulse inhibition in the CA1 region of rat hippocampal brain slices at 100 μM; induces loss of righting reflex in tadpoles (EC50 = 0.53 μM)
Catalog No. 50-293-7134 Supplier Cayman Chemical Supplier No. 287195 MG
A compound with anesthetic activity; increases paired-pulse inhibition in the CA1 region of rat hippocampal brain slices at 100 μM; induces loss of righting reflex in tadpoles (EC50 = 0.53 μM)