Learn More
Cayman Chemical Navoximod, Purity: ≥98%, CAS: 1402837-78-8, 10 mg
SDP
Catalog No. 502937953
Encompass_Preferred
An IDO1 inhibitor (IC50 = 28 nM for the recombinant human enzyme); decreases tumor volume in a Panc02 murine PDAC model at 500 mg/kg
Catalog No. 50-293-7953 Supplier Cayman Chemical Supplier No. 2844010 MG
An IDO1 inhibitor (IC50 = 28 nM for the recombinant human enzyme); decreases tumor volume in a Panc02 murine PDAC model at 500 mg/kg