Learn More
Cayman Chemical NBD-556, Purity: ≥98%, CAS: 333353-44-9, 5 mg
SDP
Catalog No. 502985453
Encompass_Preferred
A small molecule CD4 mimetic; inhibits infection of MT-2 cells by laboratory-adapted HIV-1 IIIB, MN, and V32 HIV strains (IC50 = 6.5, 15.9, and 5.3 µM, respectively), an AZT-resistant (AZT-R) HIV-1 strain (IC50 = 58 µM), and various primary HIV-1 isolates (IC50s = 15-103 μM)
Catalog No. 50-298-5453 Supplier Cayman Chemical Supplier No. 212745 MG
A small molecule CD4 mimetic; inhibits infection of MT-2 cells by laboratory-adapted HIV-1 IIIB, MN, and V32 HIV strains (IC50 = 6.5, 15.9, and 5.3 µM, respectively), an AZT-resistant (AZT-R) HIV-1 strain (IC50 = 58 µM), and various primary HIV-1 isolates (IC50s = 15-103 μM)