Learn More
Cayman Chemical NBQX, Purity: ≥98%, CAS: 118876-58-7, 5 mg
SDP
Catalog No. 502917520
Encompass_Preferred
An AMPA and kainate receptor antagonist (IC50s = 0.15 and 4.8 µM, respectively); selective for these receptors over NMDA receptors (IC50 = ≥90 µM); increases the latency to seizures, decreases the duration of seizures, and reduces seizure severity in a rat model of PTZ-induced epilepsy; suppresses relapse phase nicotine self-administration in rats
Catalog No. 50-291-7520 Supplier Cayman Chemical Supplier No. 382815 MG
An AMPA and kainate receptor antagonist (IC50s = 0.15 and 4.8 µM, respectively); selective for these receptors over NMDA receptors (IC50 = ≥90 µM); increases the latency to seizures, decreases the duration of seizures, and reduces seizure severity in a rat model of PTZ-induced epilepsy; suppresses relapse phase nicotine self-administration in rats