Learn More
Cayman Chemical NCT-501, Purity: ≥98%, CAS: 1802088-50-1, 1 mg
SDP
Catalog No. 502981087
Encompass_Preferred
A potent, reversible, and selective inhibitor of ALDH1A1 (IC50 = 40 nM for the human form)
Catalog No. 50-298-1087 Supplier Cayman Chemical Supplier No. 211621 MG
A potent, reversible, and selective inhibitor of ALDH1A1 (IC50 = 40 nM for the human form)