Learn More
Cayman Chemical ANTIBODY ND-336 500ug
SDP
Catalog No. 502982308
Encompass_Preferred
A selective inhibitor of the gelatinases MMP-2 and MMP-9, as well as MMP-14 (Kis = 85, 150, and 120 nM, respectively); accelerates diabetic wound healing in mice
Catalog No. 50-298-2308 Supplier Cayman Chemical Supplier No. 19207500 UG
A selective inhibitor of the gelatinases MMP-2 and MMP-9, as well as MMP-14 (Kis = 85, 150, and 120 nM, respectively); accelerates diabetic wound healing in mice