Learn More
Cayman Chemical ND-646, Purity: ≥98%, CAS: 1434639-57-2, 5 mg
SDP
Catalog No. 502911173
Encompass_Preferred
An ACC inhibitor (IC50s = 3.5 and 4.1 nM for human ACC1 and ACC2, respectively); inhibits production of palmitate and total fatty acid content in A549 cells at 500 nM; induces apoptosis and ER stress in A549 cells at 500 nM; inhibits fatty acid synthesis and reduces tumor growth in an A549 mouse xenograft model at 25 mg/kg twice per day
Catalog No. 50-291-1173 Supplier Cayman Chemical Supplier No. 347645 MG
An ACC inhibitor (IC50s = 3.5 and 4.1 nM for human ACC1 and ACC2, respectively); inhibits production of palmitate and total fatty acid content in A549 cells at 500 nM; induces apoptosis and ER stress in A549 cells at 500 nM; inhibits fatty acid synthesis and reduces tumor growth in an A549 mouse xenograft model at 25 mg/kg twice per day