Learn More
Cayman Chemical ANTIBODY ngenol 500ug
SDP
Catalog No. 502952872
Encompass_Preferred
A diterpenoid related to phorbol that activates PKC (Ki = 30 μM; ED50 = 27 μM) and promotes tumor formation
Catalog No. 50-295-2872 Supplier Cayman Chemical Supplier No. 14031500 UG
A diterpenoid related to phorbol that activates PKC (Ki = 30 μM; ED50 = 27 μM) and promotes tumor formation