Learn More
Cayman Chemical NIH 12848, Purity: ≥95%, CAS: 959551-10-1, 25 mg
SDP
Catalog No. 502932823
Encompass_Preferred
An inhibitor of PI5P4K (IC50 = ~1 µM); selective for PI5P4Kγ over PI5P4Kα and PI5P4Kβ (IC50s = >100 µM); inhibits translocation of the Na+/K+-ATPase to the plasma membrane in mpkCCD cells; an inhibitor of USP1/UAF complex deubiquitinase activity (IC50 = 7.9 µM); increases accumulation of monoubiquitinated PCNA in H1299 NSCLC cells at 20 µM
Catalog No. 50-293-2823 Supplier Cayman Chemical Supplier No. 2603525 MG
An inhibitor of PI5P4K (IC50 = ~1 µM); selective for PI5P4Kγ over PI5P4Kα and PI5P4Kβ (IC50s = >100 µM); inhibits translocation of the Na+/K+-ATPase to the plasma membrane in mpkCCD cells; an inhibitor of USP1/UAF complex deubiquitinase activity (IC50 = 7.9 µM); increases accumulation of monoubiquitinated PCNA in H1299 NSCLC cells at 20 µM