Learn More
Cayman Chemical ANTIBODY NIle Red 250mg
SDP
Catalog No. 502988920
Encompass_Preferred
A fluorescent probe for lipid droplets; ex/em = 529/576 nm, respectively, for neutral lipids; has been used in live, fixed, or unfixed cells; has been used as a marker of lipid droplets in cellular models of hepatocyte steatosis
Catalog No. 50-298-8920 Supplier Cayman Chemical Supplier No. 30787250 MG
A fluorescent probe for lipid droplets; ex/em = 529/576 nm, respectively, for neutral lipids; has been used in live, fixed, or unfixed cells; has been used as a marker of lipid droplets in cellular models of hepatocyte steatosis