Learn More
Cayman Chemical NMS-E973, Purity: ≥98%, CAS: 1253584-84-7, 10 mg
SDP
Catalog No. 502987958
Encompass_Preferred
An HSP90 inhibitor (IC50 = 10 nM in a fluorescence polarization assay); decreases HER2 protein expression (IC50 = 110 nM) in BT474 breast cancer cells; inhibits the growth of a 16 cancer cell line panel (IC50s = 13-362 nM); reduces tumor growth by 74% in an A2780 mouse xenograft model at a dose of 60 mg/kg, i.v., per day for ten days; completely eradicates MOLM-13 acute myeloid leukemia tumors in a mouse xenograft model
Catalog No. 50-298-7958 Supplier Cayman Chemical Supplier No. 2235210 MG
An HSP90 inhibitor (IC50 = 10 nM in a fluorescence polarization assay); decreases HER2 protein expression (IC50 = 110 nM) in BT474 breast cancer cells; inhibits the growth of a 16 cancer cell line panel (IC50s = 13-362 nM); reduces tumor growth by 74% in an A2780 mouse xenograft model at a dose of 60 mg/kg, i.v., per day for ten days; completely eradicates MOLM-13 acute myeloid leukemia tumors in a mouse xenograft model