Learn More
Cayman Chemical NMS-P118, Purity: ≥98%, CAS: 1262417-51-5, 1 mg
SDP
Catalog No. 502985260
Encompass_Preferred
A potent and selective PARP1 inhibitor; selective for PARP1 (Kd = 9 nM) over PARP2 (Kd = 1,390 nM); efficacious in mouse xenograft models of breast and pancreatic cancer
Catalog No. 50-298-5260 Supplier Cayman Chemical Supplier No. 213401 MG
A potent and selective PARP1 inhibitor; selective for PARP1 (Kd = 9 nM) over PARP2 (Kd = 1,390 nM); efficacious in mouse xenograft models of breast and pancreatic cancer