Learn More
Cayman Chemical ANTIBODY NO-Rosa5 250ug
SDP
Catalog No. 502943296
Encompass_Preferred
A photocontrollable NO donor for in cellulo and ex vivo use; composed of rosamine and N-nitrosoaminophenol; Upon excitation of rosamine between 530-590 nm, photoinduced electron transfer from the NO-releasing moiety occurs, forming an unstable phenoxy radical, which then releases NO to form a stable quinoneimine product
Catalog No. 50-294-3296 Supplier Cayman Chemical Supplier No. 42356250 UG
A photocontrollable NO donor for in cellulo and ex vivo use; composed of rosamine and N-nitrosoaminophenol; Upon excitation of rosamine between 530-590 nm, photoinduced electron transfer from the NO-releasing moiety occurs, forming an unstable phenoxy radical, which then releases NO to form a stable quinoneimine product