Learn More
Cayman Chemical ANTIBODY Norharmn 250mg
SDP
Catalog No. 502985574
Encompass_Preferred
A natural β-carboline that may be found in fried meats, cigarette smoke, and coffee; described as a co-mutagen, as it induces or enhances the mutagenicity of other compounds
Catalog No. 50-298-5574 Supplier Cayman Chemical Supplier No. 20043250 MG
A natural β-carboline that may be found in fried meats, cigarette smoke, and coffee; described as a co-mutagen, as it induces or enhances the mutagenicity of other compounds