Learn More
Cayman Chemical Nepafenac, Purity: ≥98%, CAS: 78281-72-8, 10 mg
SDP
Catalog No. 502994726
Encompass_Preferred
A prodrug from of amfenac; an inhibitor of PG synthase 1 (IC50 = 64.3 µM); inhibits the production of PGs in isolated rabbit iris/ciliary body; topical application prior to paracentesis reduces protein and PGE2 accumulation in a rabbit model of acute ocular inflammation at 0.1%
Catalog No. 50-299-4726 Supplier Cayman Chemical Supplier No. 2370010 MG
A prodrug from of amfenac; an inhibitor of PG synthase 1 (IC50 = 64.3 µM); inhibits the production of PGs in isolated rabbit iris/ciliary body; topical application prior to paracentesis reduces protein and PGE2 accumulation in a rabbit model of acute ocular inflammation at 0.1%