Learn More
Cayman Chemical NS 11021, Purity: ≥95%, CAS: 956014-19-0, 25 mg
SDP
Catalog No. 502973049
Encompass_Preferred
Selectively activates BKCa channels, binding directly the ɑ subunit; relaxes intracavernous arterial rings and corpus cavernosum strips in vitro and to enhance erectile responses in intact rats
Catalog No. 50-297-3049 Supplier Cayman Chemical Supplier No. 1534725 MG
Selectively activates BKCa channels, binding directly the ɑ subunit; relaxes intracavernous arterial rings and corpus cavernosum strips in vitro and to enhance erectile responses in intact rats