Learn More
Cayman Chemical NSC 109555, Purity: ≥98%, CAS: 15427-93-7, 5 mg
SDP
Catalog No. 502984497
Encompass_Preferred
An inhibitor of Chk2 (IC50 = 200 nM in a cell-free kinase assay); selective for Chk2 over Chk1 and 16 kinases in a panel but does inhibit Brk, c-Met, IGFR, and LCK (IC50s = 210, 6,000, 7,400, and 7,100 nM, respectively); inhibits Chk2 autophosphorylation and phosphorylation of the Chk2 substrate histone H1 in vitro (IC50 = 240 nM); inhibits the growth of, and induces autophagy in, L1210 leukemia cells in vitro; potentiates gemcitabine-induced cytotoxicity in MIA PaCa-2, CFPAC-1, PANC-1, and BxPC-3 pancreatic cancer cells, as well as reduces gemcitabine-induced increases in Chk2 phosphorylation and enhances gemcitabine-induced production of ROS in MIA PaCa-2 cells at 1,250 nM
Catalog No. 50-298-4497 Supplier Cayman Chemical Supplier No. 198115 MG
An inhibitor of Chk2 (IC50 = 200 nM in a cell-free kinase assay); selective for Chk2 over Chk1 and 16 kinases in a panel but does inhibit Brk, c-Met, IGFR, and LCK (IC50s = 210, 6,000, 7,400, and 7,100 nM, respectively); inhibits Chk2 autophosphorylation and phosphorylation of the Chk2 substrate histone H1 in vitro (IC50 = 240 nM); inhibits the growth of, and induces autophagy in, L1210 leukemia cells in vitro; potentiates gemcitabine-induced cytotoxicity in MIA PaCa-2, CFPAC-1, PANC-1, and BxPC-3 pancreatic cancer cells, as well as reduces gemcitabine-induced increases in Chk2 phosphorylation and enhances gemcitabine-induced production of ROS in MIA PaCa-2 cells at 1,250 nM