Learn More
Cayman Chemical NSC 44625, Purity: ≥98%, CAS: 654-62-6, 1 g
SDP
Catalog No. 502943791
Encompass_Preferred
A CAII inhibitor (Ki = 63 nM for the human enzyme); selective for CAII over CAI (Ki = 5,800 nM for the human enzyme) but also inhibits β-carbonic anhydrase (Ki = 223 nM for the S. cerevisiae enzyme)
Catalog No. 50-294-3791 Supplier Cayman Chemical Supplier No. 409851 G
A CAII inhibitor (Ki = 63 nM for the human enzyme); selective for CAII over CAI (Ki = 5,800 nM for the human enzyme) but also inhibits β-carbonic anhydrase (Ki = 223 nM for the S. cerevisiae enzyme)