Learn More
Cayman Chemical NSC 668036, Purity: ≥98%, CAS: 144678-63-7, 10 mg
SDP
Catalog No. 502964505
Encompass_Preferred
An inhibitor of the Dvl PDZ domain (Kd = 240 μM for mouse Dvl1 PDZ domain); reduces expression of the Wnt target gene, Siamois, in Xenopus embryos and inhibits the ability of injected Wnt3A to induce formation of a secondary axis (180 ng); inhibits proliferation of NIH/3T3 fibroblasts, restrains their migration in a scratch assay, and blocks the accelerating effect of TGF-β on fibroblast migration (>10 μM); reduces the severity of bleomycin-induced pulmonary fibrosis and expression of α-SMA collagen I, TGF-β, and β-catenin in lung in mice (5 mg/kg per day for 14 days)
Catalog No. 50-296-4505 Supplier Cayman Chemical Supplier No. 2349910 MG
An inhibitor of the Dvl PDZ domain (Kd = 240 μM for mouse Dvl1 PDZ domain); reduces expression of the Wnt target gene, Siamois, in Xenopus embryos and inhibits the ability of injected Wnt3A to induce formation of a secondary axis (180 ng); inhibits proliferation of NIH/3T3 fibroblasts, restrains their migration in a scratch assay, and blocks the accelerating effect of TGF-β on fibroblast migration (>10 μM); reduces the severity of bleomycin-induced pulmonary fibrosis and expression of α-SMA collagen I, TGF-β, and β-catenin in lung in mice (5 mg/kg per day for 14 days)