Learn More
Cayman Chemical NSC 668394, Purity: ≥98%, CAS: 382605-72-3, 25 mg
SDP
Catalog No. 502980068
Encompass_Preferred
An ezrin binder (Kd = 10.6 µM); inhibits PKCι-induced phosphorylation of ezrin in a cell-free assay (IC50 = 8.1 µM); inhibits the interaction between ezrin and actin in K7M2 cells at 10 µM; reduces bacterial invasion but is not bactericidal in HEp-2 cells cells infected with GAS at 10 µM; induces developmental delays and cyclopia in Xenopus larvae at 10 µM and induces toxicity at 40 µM
Catalog No. 50-298-0068 Supplier Cayman Chemical Supplier No. 1686325 MG
An ezrin binder (Kd = 10.6 µM); inhibits PKCι-induced phosphorylation of ezrin in a cell-free assay (IC50 = 8.1 µM); inhibits the interaction between ezrin and actin in K7M2 cells at 10 µM; reduces bacterial invasion but is not bactericidal in HEp-2 cells cells infected with GAS at 10 µM; induces developmental delays and cyclopia in Xenopus larvae at 10 µM and induces toxicity at 40 µM