Learn More
Cayman Chemical NSC 756093, Purity: ≥98%, CAS: 1629908-92-4, 10 mg
SDP
Catalog No. 502912301
Encompass_Preferred
An inhibitor of the GBP1-Pim-1 protein-protein interaction; binds to GBP1-Pim-1 (Kd = 38 nM) and inhibits the interaction by 65% at 100 nM; enhances radiation-induced cytotoxicity in FaDu cells (IC50 = 496 nM); induces apoptosis and cell cycle arrest at the G2/M phase in FaDu cells
Catalog No. 50-291-2301 Supplier Cayman Chemical Supplier No. 3502310 MG
An inhibitor of the GBP1-Pim-1 protein-protein interaction; binds to GBP1-Pim-1 (Kd = 38 nM) and inhibits the interaction by 65% at 100 nM; enhances radiation-induced cytotoxicity in FaDu cells (IC50 = 496 nM); induces apoptosis and cell cycle arrest at the G2/M phase in FaDu cells