Learn More
Cayman Chemical NSC 781406, Purity: ≥98%, CAS: 1676893-24-5, 1 mg
SDP
Catalog No. 502938119
Encompass_Preferred
A dual inhibitor of PI3K and mTOR (IC50s = 2.0, 9.4, 2.7, 14, and 5.4 nM for PI3Kα, PI3Kβ, PI3Kγ, PI3Kδ, and mTOR, respectively); inhibits cell growth in the NCI-60 panel of cancer cell lines (mean GI50 = 65 nM); reduces tumor volume in a BEL-7404 hepatic cancer mouse xenograft model at 30 mg/kg
Catalog No. 50-293-8119 Supplier Cayman Chemical Supplier No. 292261 MG
A dual inhibitor of PI3K and mTOR (IC50s = 2.0, 9.4, 2.7, 14, and 5.4 nM for PI3Kα, PI3Kβ, PI3Kγ, PI3Kδ, and mTOR, respectively); inhibits cell growth in the NCI-60 panel of cancer cell lines (mean GI50 = 65 nM); reduces tumor volume in a BEL-7404 hepatic cancer mouse xenograft model at 30 mg/kg