Learn More
Cayman Chemical NSC 87877, Purity: ≥98%, CAS: 56990-57-9, 50 mg
SDP
Catalog No. 502973046
Encompass_Preferred
A cell-permeable, potent inhibitor of both SHP-1 and SHP-2 (IC50 = 355 and 318 nM, respectively); much less effective against other protein tyrosine phosphatases and DUSP26
Catalog No. 50-297-3046 Supplier Cayman Chemical Supplier No. 1490850 MG
A cell-permeable, potent inhibitor of both SHP-1 and SHP-2 (IC50 = 355 and 318 nM, respectively); much less effective against other protein tyrosine phosphatases and DUSP26