Learn More
Cayman Chemical NSC 95397, Purity: ≥98%, CAS: 93718-83-3, 10 mg
SDP
Catalog No. 502994017
Encompass_Preferred
A selective, irreversible inhibitor of Cdc25 dual-specificity phosphatases, with Ki values of 32, 96, and 40 nM for Cdc25A, Cdc25B, and Cdc25C, respectively; significantly inhibits the growth of human and murine carcinoma cells, blocking G2/M phase transition
Catalog No. 50-299-4017 Supplier Cayman Chemical Supplier No. 2143110 MG
A selective, irreversible inhibitor of Cdc25 dual-specificity phosphatases, with Ki values of 32, 96, and 40 nM for Cdc25A, Cdc25B, and Cdc25C, respectively; significantly inhibits the growth of human and murine carcinoma cells, blocking G2/M phase transition