Learn More
Cayman Chemical Nevirapine, Purity: ≥98%, CAS: 129618-40-2, 25 mg
SDP
Catalog No. 502971467
Encompass_Preferred
An NNRTI; binds to HIV-1 reverse transcriptase and inhibits RNA plus-strand initiation (IC50 = 0.45 μM); prevents seroconversion and viremia in a chimpanzee model of HIV-1 infection
Catalog No. 50-297-1467 Supplier Cayman Chemical Supplier No. 1511725 MG
An NNRTI; binds to HIV-1 reverse transcriptase and inhibits RNA plus-strand initiation (IC50 = 0.45 μM); prevents seroconversion and viremia in a chimpanzee model of HIV-1 infection