Learn More
Cayman Chemical NVP-ADW742, Purity: ≥98%, CAS: 475488-23-4, 1 mg
SDP
Catalog No. 502980073
Encompass_Preferred
A selective IGF-1R inhibitor (IC50 = 0.17 µM); inhibits tumor proliferation both in vitro and in a mouse model of multiple myeloma
Catalog No. 50-298-0073 Supplier Cayman Chemical Supplier No. 183901 MG
A selective IGF-1R inhibitor (IC50 = 0.17 µM); inhibits tumor proliferation both in vitro and in a mouse model of multiple myeloma