Learn More
Cayman Chemical NVP-BAG956, Purity: ≥98%, CAS: 853910-02-8, 5 mg
SDP
Catalog No. 502935945
Encompass_Preferred
A dual PDK1 and class I PI3K inhibitor (IC50s = 245, 56, 446, 35, and 117 nM for PDK1, p110α, p110β, p110δ, and p110γ, respectively); inhibits phosphorylation of T308-PKB (IC50 = 45 nM) in and proliferation of U87MG cells (EC50 = 143 nM); induces cell cycle arrest at the G1 phase in MOLM-14 cells and reduces colony formation by patient-derived AML cells; reduces tumor burden in mice injected with 32D.p210-luc+cells when administered in combination with everolimus
Catalog No. 50-293-5945 Supplier Cayman Chemical Supplier No. 241955 MG
A dual PDK1 and class I PI3K inhibitor (IC50s = 245, 56, 446, 35, and 117 nM for PDK1, p110α, p110β, p110δ, and p110γ, respectively); inhibits phosphorylation of T308-PKB (IC50 = 45 nM) in and proliferation of U87MG cells (EC50 = 143 nM); induces cell cycle arrest at the G1 phase in MOLM-14 cells and reduces colony formation by patient-derived AML cells; reduces tumor burden in mice injected with 32D.p210-luc+cells when administered in combination with everolimus