Learn More
Cayman Chemical ANTIBODY NVP-BEP800 500ug
SDP
Catalog No. 502967601
Encompass_Preferred
An Hsp90 inhibitor (IC50 = 58 nM for recombinant human Hsp90β); selective for Hsp90 over GRP94 and TRAP1 (IC50s = 4.1 and 5.5 µM, respectively); inhibits the growth of BT474, SK-BR-3, MCF-7, MDA-MB-157, MDA-MB-231, MDA-MB-468, and BT-20 breast cancer cells (GI50s = 53, 56, 118, 89, 190 173, and 162 nM, respectively); reduces levels of HER2 and Raf-1 in MCF-7 cells; induces apoptosis and attenuates X-ray-induced increases in Hsp70 levels in T98G glioblastoma cells; reduces tumor volume in A375 melanoma and BT474 breast cancer mouse xenograft models from 15-50 mg/kg; reduces tumor volume in an L1T2 Kaposi sarcoma mouse xenograft model
Catalog No. 50-296-7601 Supplier Cayman Chemical Supplier No. 18383500 UG
An Hsp90 inhibitor (IC50 = 58 nM for recombinant human Hsp90β); selective for Hsp90 over GRP94 and TRAP1 (IC50s = 4.1 and 5.5 µM, respectively); inhibits the growth of BT474, SK-BR-3, MCF-7, MDA-MB-157, MDA-MB-231, MDA-MB-468, and BT-20 breast cancer cells (GI50s = 53, 56, 118, 89, 190 173, and 162 nM, respectively); reduces levels of HER2 and Raf-1 in MCF-7 cells; induces apoptosis and attenuates X-ray-induced increases in Hsp70 levels in T98G glioblastoma cells; reduces tumor volume in A375 melanoma and BT474 breast cancer mouse xenograft models from 15-50 mg/kg; reduces tumor volume in an L1T2 Kaposi sarcoma mouse xenograft model