Learn More
Cayman Chemical NVS-PAK1-1, Purity: ≥98%, CAS: 1783816-74-9, 10 mg
SDP
Catalog No. 502983028
Encompass_Preferred
An allosteric inhibitor of PAK1 (Kd = 7 nM); has an IC50 value of 5.2 nM in a PAK1 dephosphorylation assay; selective for PAK1 over PAK2 (Kd = 400 nM) and over a panel of 442 kinases; inhibits phosphorylation of MEK Ser289 at 6-20 µM but does not inhibit proliferation of Su86.86 cells at concentrations lower than 2 µM
Catalog No. 50-298-3028 Supplier Cayman Chemical Supplier No. 1996410 MG
An allosteric inhibitor of PAK1 (Kd = 7 nM); has an IC50 value of 5.2 nM in a PAK1 dephosphorylation assay; selective for PAK1 over PAK2 (Kd = 400 nM) and over a panel of 442 kinases; inhibits phosphorylation of MEK Ser289 at 6-20 µM but does not inhibit proliferation of Su86.86 cells at concentrations lower than 2 µM