Learn More
Cayman Chemical o,p'-DDE, Purity: ≥95%, CAS: 3424-82-6, 10 mg
SDP
Catalog No. 502935485
Encompass_Preferred
A metabolite and degradation product of the pesticide DDT; accumulates in various fish and sediment surrounding a DDT manufacturing plant; inhibits estrogen binding to rtERs (IC50 = 3.2 μM); induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; increases degeneration of ovarian follicles and reduces testicular size in Japanese medaka (O. latipes)
Catalog No. 50-293-5485 Supplier Cayman Chemical Supplier No. 2423510 MG
A metabolite and degradation product of the pesticide DDT; accumulates in various fish and sediment surrounding a DDT manufacturing plant; inhibits estrogen binding to rtERs (IC50 = 3.2 μM); induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; increases degeneration of ovarian follicles and reduces testicular size in Japanese medaka (O. latipes)